L(-)-Fucose, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALF2252-100GEA 3280 EUR
SIALF2252-100G SIALF2252-10MG SIALF2252-25G SIALF2252-5G
L(-)-Fucose, Sigma-Aldrich®
L(-)-Fucose
Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
MDL: MFCD00069812
CAS-Nummer: 2438-80-4
EINECS: 219-452-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR