Natriumhexanoat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIALC4026-25GEA 204 EUR
SIALC4026-25G SIALC4026-10G
Natriumhexanoat, Sigma-Aldrich®
Natriumhexanoat
Formel: C₆H₁₁NaO₂
MDL: MFCD00059056
CAS-Nummer: 10051-44-2
EINECS: 233-179-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR