1,2-Epoxycycloheptan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL672440-5GEA 585 EUR
SIAL672440-5G SIAL672440-1G
1,2-Epoxycycloheptan, Sigma-Aldrich®
1,2-Epoxycycloheptane
Formel: C₇H₁₂O
MDL: MFCD00022357
CAS-Nummer: 286-45-3
EINECS: 206-009-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR