In-line filters, SSI™
Lieferant: Merck
SIAL59262-UEA 170 EUR
SIAL59262-U SIAL59264 SIAL59265
In-line filters, SSI™
Filter In-Line Filter
The high pressure in-line filter should be place between the pump and injector to provide final filtration for the mobile phase.
Maximum operating pressure: 1034,21 bar