Benzophenon-[D10] (99% D), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL471178-5GEA 871 EUR
SIAL471178-5G SIAL471178-1G
Benzophenon-[D10] (99% D), Sigma-Aldrich®
Benzophenon-[D10]
Formel: C₁₃D₁₀O
CAS-Nummer: 22583-75-1
EINECS: 245-107-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR