3-(Chlormethyl)benzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL424315-5GEA 42.7 EUR
SIAL424315-5G SIAL424315-25G
3-(Chlormethyl)benzoesäure, Sigma-Aldrich®
3-(Chloromethyl)benzoic acid
Formel: C₈H₇ClO₂
Molecular Weight: 170,59 g/mol
MDL: MFCD00191922
CAS-Nummer: 31719-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR