(R)-(-)-Methyl-3-hydroxyisobutyrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL270148-5GEA 176 EUR
SIAL270148-5G SIAL270148-1G
(R)-(-)-Methyl-3-hydroxyisobutyrat, Sigma-Aldrich®
(R)-(-)-Methyl-3-hydroxyisobutyrat
Formel: C₅H₁₀O₃
Molecular Weight: 118,1311 g/mol
MDL: MFCD00063450
CAS-Nummer: 72657-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR