Di(2-pyridyl)keton, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL127221-25GEA 494 EUR
SIAL127221-25G SIAL127221-5G
Di(2-pyridyl)keton, Sigma-Aldrich®
Di(2-pyridyl)keton
Formel: C₁₁H₈N₂O
Molecular Weight: 184,20
MDL: MFCD00006288
CAS-Nummer: 19437-26-4
EINECS: 243-067-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR