Safety disposal unit (funnel-model) with drain tray
Lieferant: Semadeni
SEMA4500EA 325 EUR
SEMA4500 SEMA4502 SEMA4501 SEMA4503
Safety disposal unit (funnel-model) with drain tray
Sicherheitsbehälter
Safety disposal canister in a drain tray made of polypropylene (PP) for the disposal of hazardous waste.
  • With a screw-on funnel
  • Inward sieve and level indicator to avoid overfilling
  • Large opening, self closing lid to prevent odour and corrosion in the room