Sample tubes
Lieferant: Semadeni
SEMA3197EA 39.5 EUR
SEMA3197 SEMA3199
Sample tubes
Röhrchen Reagenzgläser- und Probenröhrchen
PP, with screw cap.
  • Excellent chemical resistance
  • Excellent temperature resistance