Tubing clamps, POM
Lieferant: Semadeni
SEMA2492EA 94.8 EUR
SEMA2492
Tubing clamps, POM
Klemmen Schlauchklemmen und Sanitärklemmen
Polyacetal (POM), blue. Suitable for catheters, tubing and pipes up to 10 mm diameter.
  • Solid and sterilisable
  • Quick and easy opening and closing
  • Temperature resistant up to +130 °C
Suitable for catheters, tubing and pipes up to 10 mm diameter.
  • Solid and sterilisable
  • Quick and easy opening and closing
  • Temperature resistant up to +130 °C