Multipurpose basins, with handles
Lieferant: Semadeni
SEMA1752EA 21.8 EUR
SEMA1752
Multipurpose basins, with handles
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
PP, granite.
  • Rectangular, with two side handles
  • Stackable and resistant in hot water
  • Suitable for food
Multipurpose basins for a wide range of applications.