Transport tubes, with stopper
Lieferant: Semadeni
SEMA1699EA 32.3 EUR
SEMA1699 SEMA1698 212-3232
Transport tubes, with stopper
Röhrchen Reagenzgläser- und Probenröhrchen
PS, clear, with white push-in stopper.
  • Flat bottom
  • Good thermal resistance
  • Good chemical resistance
Universal tubes for a wide range of applications.