Kidney dishes
Lieferant: Semadeni
SEMA1254EA 21.8 EUR
SEMA1254
Kidney dishes
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Kidney dishes made of white PP.
  • Temperature resistant up to 130 °C
  • Excellent chemical resistance