Clamps
Lieferant: Semadeni
SEMA1082EA 4.1 EUR
SEMA1082 SEMA1081 SEMA1080 SEMA1078 SEMA1077 SEMA1076 SEMA1086 SEMA1075 SEMA1084
Clamps
Klemmen Klemmen für den allgemeinen Gebrauch
Clamps made of polyacetal (POM), in light blue.
  • Chemical and temperature resistant
  • Sticks (80 mm high) can be cut to the desired height