Tubing connectors, T-shaped
Lieferant: Semadeni
SEMA0469EA 3 EUR
SEMA0469 SEMA0467 SEMA0392
Tubing connectors, T-shaped
Fittings und Verbinder
PP, translucent.
  • Good mechanical and chemical resistance
  • Very good thermal resistance
  • Autoclavable
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine "grip". This will then give equivalent crest dimension for the particular connector concerned.