Magnetic stirring bars, double-ended
Lieferant: Semadeni
SEMA0332EA 24.8 EUR
SEMA0332 SEMA0331
Magnetic stirring bars, double-ended
Rührer Magnetrührstäbchen
Double ended magnetic stirring bars provide excellent centering, limited contact surface and high turbulence even at low speeds.
  • Ideal for test tubes or for eccentric stirring movements