Magnetic stirring bars, cylindrical
Lieferant: Semadeni
SEMA0259EA 11.2 EUR
SEMA0259 SEMA0258 SEMA0257 SEMA0268 SEMA0244 SEMA0263_U SEMA0320 SEMA2209
Magnetic stirring bars, cylindrical
Rührer Magnetrührstäbchen
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.
  • Chemical resistance
  • Smooth surface and round ends
  • Temperature resistance upto 275 °C