Sicherheitstrichter mit Deckel
Lieferant: S.C.A.T.
SCTV107670EA 189 EUR
SCTV107670 SCTV108670
Sicherheitstrichter mit Deckel
Trichter Sicherheitstrichter
Safety funnels with hinged lid cap, sieve and safety lance.
  • Lid funnel with double thread R2" BSP/G2" (m) + 2" Tri-Sure (m) or S60/61
  • Made of PE-HD material
  • Stable stainless-steel strainer
  • Large funnel diameter: 250 mm
The funnels with hinged lid are suitable for universal use in the laboratory and are highly resistant to conventional chemicals due to the high-quality PE-Hd material.