Multiport connector caps, GL 25, DURAN®
Lieferant: DWK Life Sciences
DURAN®
SCOT292611302EA 61.67 EUR
SCOT292611302 SCOT292621303 SCOT292601301
Multiport connector caps, GL 25, DURAN®
Fittings und Verbinder
DURAN® GL 25 connector caps with multiple ports are suitable for a range of applications were small volumes of liquid need to be sampled or dispensed.
  • High temperature resistance (up to 180 °C)
  • Caps are fully autoclavable and reusable
The barbed male tubing connectors are made from a surgical grade stainless steel (316L) and will fit tubing with a Ø 3,2 mm.