Standard DG-geschliffen
Lieferant: DWK Life Sciences
SCOT286518001EA 115 EUR
SCOT286518001 SCOT286154208 SCOT286521003 SCOT286541202 SCOT286156303 SCOT286158801 SCOT286155401 SCOT286532806 SCOT286546003 SCOT286558202
Standard DG-geschliffen
Adapter Übergangsstücke mit Kegelschliffen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With Diamond ground.