Exsikkator, MOBILEX, mit Porzellanplatte DURAN®
Lieferant: DWK Life Sciences
DURAN®
SCOT247836955EA 790.26 EUR
SCOT247836955 SCOT247836655 SCOT247835753 SCOT247836158
Exsikkator, MOBILEX, mit Porzellanplatte DURAN®
Exsikkatoren Exsikkatoren, Glas
DURAN® clear borosilicate glass 3.3, vacuum desiccator with Mobilex screw thread in the lid with stopcock. With desiccator base with flat flange, no outlet, PBT screw cap with pierced aperture GL 32.
Lieferumfang: Supplied with porclain plate and silicone sealing ring with bonded PTFE face