Automatic Burettes, Pellet Pattern, Class AS
Lieferant: DWK Life Sciences
SCOT243172704EA 297.2 EUR
SCOT243172704 SCOT243173606 SCOT243173306 SCOT243173355 SCOT243173655 SCOT243172753
Automatic Burettes, Pellet Pattern, Class AS
Büretten
Burette, automatic, Pellet, DURAN® borosilicate glass, with PTFE key, class AS.
  • 30 seconds waiting time
  • Schellbach stripe and main graduations as circular divisions
  • Supplied with reservoir bottle and blow ball
Standards: DIN EN ISO 385