Measuring cylinders, SUPER DUTY, borosilicate glass, low form, class B, DURAN®
Lieferant: DWK Life Sciences
DURAN®
SCOT213942406EA 95.46 EUR
SCOT213942406 SCOT213944407 SCOT213943608 SCOT213945403
Measuring cylinders, SUPER DUTY, borosilicate glass, low form, class B, DURAN®
Zylinder Messzylinder
DURAN®, borosilicate glass, class B, low form.
  • Very good chemical resistance, high temperature resistance and minimal thermal expansion
  • White easy-to-read scale, highly durable white ceramic
  • Economical
  • Increased safety due to lower breakage risk
  • Supplied with batch certificate
Ideal for demanding applications such as field operations, academic and professional training and frequent stress due to automated washing.
Standards: USP Standard; ISO 4788; with printed retrace code (batch identification).