ORP electrode solutions
Lieferant: SI Analytics
SCOI285138373EA 178 EUR
SCOI285138373 662-6171 SCOI285138381 SCOI285138357 662-1609 SCOI285138168
ORP electrode solutions
Standards Redox-/ORP-Standards
ORP electrode solutions are used as cleaning solutions for combination electrodes and reference electrodes.