Couch cover rolls, Tork
Lieferant: Essity
Tork® Tork® Advanced Tork® Universal
SCAA125149EA 129 EUR
SCAA125149 115-3115 SCAA125250 SCAA125146
Couch cover rolls, Tork
Oberflächenschutz Möbelauflagen
Hygienic couch cover rolls for protection of patient couches made from strong fibres.
  • Perforated to reduce consumption
  • Tork Universal made from recycled fibres
  • Tork Advanced made from virgin fibres
For use with Tork couch roll dispensers.
Bestellinformation: Available in different materials, one or two ply in multiple lengths and widths.