Funnels, disposable, Microsart® 100/250
Lieferant: Sartorius
Microsart®
514-0190EA 179 EUR
514-0190 SART16A07-10-N
Funnels, disposable, Microsart® 100/250
Trichter Membranfiltertrichter
PP housing.
Microsart® funnels are sterile 100 ml and 250 ml respectively plastic funnels. They allow quick performance of the filtrations required in the routine testing of water, food and beverages, pharmaceutical and cosmetic products.
  • Reliable results-use a new, sterile funnel for each test for certain avoidances of cross contamination
  • Time saving-kust change the funnel, rather than spending time sanitizing it
  • Simpler handling-no more holding of hot funnels. And you can see when filtration has been completed, particulary useful when using manifolds in routine testing