Mini-Drums and light duty drums (No toogles)
Lieferant: Sampling Systems
SAMPA446-10EA 1350 EUR
SAMPA446-10 SAMPA466-30 SAMPA466-20 SAMPA466-50 SAMPA446-15 SAMPA466-25 SAMPA446-12 SAMPA446-5 SAMPA454-30 SAMPA446-2 SAMPA446-1 SAMPA454-20 SAMPA454-25
Mini-Drums and light duty drums (No toogles)
Fässer, Fässer, Trommeln
Made of stainless steel.
  • Light duty construction
  • With crevice free interior
  • Flat bottom with chime ring to give added protection
  • With lid
Multi purpose containers.
Bitte beachten If you need a watertight container please ask for drum with toggles.