Pulvertrichter
Lieferant: Sampling Systems
SAMP8023P-100EA 14.4 EUR
SAMP8023P-100 SAMP8023P-350 SAMP8023P-250 SAMP8023P-150
Pulvertrichter
Trichter Pulvertrichter
Reusable powder funnel, made of PP, crevice free, with wide diameter discharge spout to enable powder to be discharged without catching.
  • Good chemical resistance
  • Good temperature resistance
  • Ideal for loading powder into drums, containers or kegs