Tiegelzangen, doppelt gebogen
Lieferant: RSG Solingen
RSGA081.300EA 30.7 EUR
RSGA081.300 RSGA081.220 RSGA081.250 RSGA081.200
Tiegelzangen, doppelt gebogen
Zangen
These scissor-type crucible tongs are made of electropolished stainless steel. With double curved tips.