High pressure frit-type in-line filter
Lieferant: Restek
REST25041EA 81 EUR
REST25041
High pressure frit-type in-line filter
Filter In-Line Filter
This high pressure in-line filter is a stand-alone version of the Trident column protection system, specifically designed for ease of use, low dead-volume, and flexibility.
  • Use of this filter can greatly extend column life
The filter has a replaceable, PEEK encapsulated 316 stainless steel frit with a surface area of 12 mm². The standard frit with 2,0 μm porosity may be replaced with an optional 0,5 μm porosity frit.