In-line filters, high-pressure cup-type
Lieferant: Restek
REST25001EA 0 EUR
REST25001 REST25002 REST25000 REST25003
In-line filters, high-pressure cup-type
Filter In-Line Filter
High-pressure cup-type filter made up of 316 stainless steel which is used in fluid streams operating to 1034 bar.
  • Large surface area (2,5 cm²) gives long operating lifetime
  • Mounted in screw-type adapters
Tubing: ¹/₁₆" Ext.Ø
Lieferumfang: Filters are supplied with gland nuts and ferrules.