Osmolality standards provide ease of use when calibrating all types of osmometers.
- Ready to use and presented in convenient ampoules
- Extended shelf life and has low uncertainty of measurement
- Consistency of product-Independent, traceable
- Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Standards: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.