Osmolality standards
Lieferant: Reagecon
REGERE-OSM-50EA 169 EUR
REGERE-OSM-50 REGERE-OSM-1500 REGERE-OSM-290 REGERE-OSM-850 REGERE-OSM-300 REGERE-OSM-400 REGERE-OSM-100 REGERE-OSM-200 REGERE-OSM-1000 REGERE-OSM-500 REGERE-OSM-900 REGERE-OSM-3000 REGERE-OSM-2000
Osmolality standards
Standards Osmolalitäts-Standards
Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.
  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml
The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.
Standards: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.