Chemical resistant overalls, BizTex® Microporous, ST60
Lieferant: Portwest
BizTex®
PRTWST60WHRXXLEA 519 EUR
PRTWST60WHRXXL PRTWST60WHRM PRTWST60WHRL PRTWST60WHRXL
Chemical resistant overalls, BizTex® Microporous, ST60
Einweg-Kleidung
This overall made of BizTex® Microporous fabric offers excellent chemical and dust protection from biological agents.
  • Ultra low linting with bound tape seams: extremely effective barrier to chemicals
  • Ergonomically design for unrestricted movement comfort and breathability
  • Elasticated 3-piece hood with chinflap
Weight: 60 g/m²
Standards: EN 14605 (Type 4), EN 13034 (type 6), EN ISO 13982 (type 5), EN 1149-5, EN 1073-2, EN 14126, Cat. III