Shoe covers, BizTex® Microporous type PB[6]
Lieferant: Portwest
BizTex®
PRTWST44WHREA 185 EUR
PRTWST44WHR
Shoe covers, BizTex® Microporous type PB[6]
Einweg-Kleidung
These protective overshoes are designed for use in all work environments where shoes or floors need protecting. Elastic finish to top of shoe ensures a secure fit.
  • Made of microporous polypropylene/polyethylene
Standards: This fabric conforms to EN 14126 biological protection and EN 1149-1 anti-static protection, EN 13034 Type PB [6].
Bitte beachten Do not iron and wash.