Overalls, PP, ST11
Lieferant: Portwest
PRTWST11NARXXXLEA 0 EUR
PRTWST11NARXXXL PRTWST11WHRXXL PRTWST11NARXL PRTWST11WHRXL PRTWST11WHRL PRTWST11WHRXXXL PRTWST11WHRM PRTWST11NARXXL PRTWST11NARM PRTWST11WHRS PRTWST11NARL
Overalls, PP, ST11
Einweg-Kleidung
This disposable lightweight overall are made from a breathable non woven PP fabric, enabling the user to keep cool while wearing the garment.
  • Generous size design provides exceptional comfort
  • Shields against non-hazardous dirt, dust and grime
Weight: 40 g