Chemical kit, 50 L, PW® spill, SM91
Lieferant: Portwest
PRTWSM91YEREA 236 EUR
PRTWSM91YER
Chemical kit, 50 L, PW® spill, SM91
Absorptionsmaterial
This kit can be used in exterior and interior environments. Reinforced construction of pads and socks allow retention of harmful substances such as acids and solvents.
Standards: BS 7959-1, BS 7959-3.