Chemical socks, PW® spill, SM70
Lieferant: Portwest
PRTWSM70YEREA 165 EUR
PRTWSM70YER
Chemical socks, PW® spill, SM70
Absorptionsmaterial
The chemical sock is perfect for environments where fast, effective containment and control of hazardous and non-hazardous chemicals is essential. Flexible fabric expands rapidly to increase the rate of absorption while providing excellent liquid retention, durability and strength.
  • Polyester knitted fabric outer with polypropylene fill
Standards: BS 7959-1, BS 7959-3.