Blades for safety cutter KN40
Lieferant: Portwest
PRTWKN91NCREA 89.9 EUR
PRTWKN91NCR
Blades for safety cutter KN40
Rasiermesser und Klingen
These blades are constructed from carbon steel.
  • Each blade features two cutting edges
Designed to be used with the KN40 safety cutter.