Elemental analysis standards, sulphur standards, VHG
Lieferant: LGC Standards
VHG
PROOVHGSDSL2P100EA 133 EUR
PROOVHGSDSL2P100 PROOVHGSDSL5000100 PROOVHGSDSL300100 PROOVHGSDSL75100 PROOVHGSDSLBLK100 PROOVHGSDSL1000100 PROOVHGSDSL4P100 PROOVHGSDSL50100 PROOVHGSDSL15100 PROOVHGSDSL750100 PROOVHGSDSL10100 PROOVHGSDSL3P100 PROOVHGSDSL100100 PROOVHGSDSL400100 PROOVHGSDSL1500100 PROOVHGSDSL500100 PROOVHGSDSL5P100 PROOVHGSDSL3000100 PROOVHGSDSL7500100 PROOVHGSDSL25100 PROOVHGSDSL1P100 PROOVHGSDSL20100 PROOVHGSDSL200100 PROOVHGSDSL5100
Elemental analysis standards, sulphur standards, VHG
Standards Kationischer Standards Schwefel-Standards
This standard solution is intended for use as a certified reference material and calibration standard for the analysis of sulphur in various petroleum products using x-ray fluorescence spectrometry (XRF), combustion and other techniques for sulphur analysis. It is suitable for use with methods such as ASTM D2622, D4294, D5453, D7039, D7212, D7220 and others.
Standards: Traceable to NIST and SRM.