Reference materials, acid number standards, in hydrocarbon oil, VHG
Lieferant: LGC Standards
PROOVHG-AN-1-100GEA 214 EUR
PROOVHG-AN-1-100G PROOVHGAN-0.5-100G PROOVHGAN-0.1-100G PROOVHGAN-1.5-100G
Reference materials, acid number standards, in hydrocarbon oil, VHG
Standards Mineralölindustrie-Standards
These reference materials are intended for use in the determination of acid number (AN) in petroleum products in accordance with ASTM D664/IP 177 (by potentiometric titration) or ASTM D974 (by colour-indicator titration).
Lieferumfang: Each standard is supplied with a full certificate of analysis (COA) which states the certified values and uncertainty by both ASTM D664 and D974.