Viscosity standards, VHG
Lieferant: LGC Standards
VHG
PROOVHG-VISC120EA 107 EUR
PROOVHG-VISC120 PROOVHG-VISC10-500 PROOVHG-VISC30-500 PROOVHG-VISC5-500 PROOVHG-VISC60A PROOVHG-VISC20-500 PROOVHG-VISC50-500 PROOVHG-VISC900
Viscosity standards, VHG
Standards Viskosimetrie-Standards
These standard solutions are intended for use as a certified reference materiasl and calibration standards for viscometric instrumentation, using ASTM Method D445 and other viscometric methods.
Standards: Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST.