Multi-element metallo-organic standards, V21 wear metals standard, VHG
Lieferant: LGC Standards
VHG
PROOVHG-V21-50-200EA 152 EUR
PROOVHG-V21-50-200 PROOVHG-V21-50-100 PROOVHG-V21100-200 PROOVHG-V21-10-200 PROOVHG-V21100-100 PROOVHG-V21-10-100 PROOVHG-V21-30-100 PROOVHG-V21300-100 PROOVHG-V21300-200 PROOVHG-V21-30-200
Multi-element metallo-organic standards, V21 wear metals standard, VHG
Standards Kationischer Standards Mehrelement-Standards
These standards are intended for use as a certified reference material (CRM) and calibration standard for the analysis of these elements in petroleum products and other organic matrices using inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), rotating disc electrode atomic emission spectroscopy (RDE-AES), flame or furnace atomic absorption spectroscopy (AA or GFAA) and other techniques for elemental analysis.
Multi element standards for Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in hydrocarbon oil.
Standards: Accredited to ISO 9001, ISO Guide 34 and ISO/IEC 17025. Traceable to NIST and SRM.