Strontium standard solution, for ICP, in HNO₃, CRM, VHG
Lieferant: LGC Standards
PROOVHG-TSRN-100EA 92 EUR
PROOVHG-TSRN-100 PROOVHG-TSRN-500 PROOVHG-PSRN-500 PROOVHG-PSRN-100 PROOVHG-PSRN-50 PROOVHG-LSRN-100
Strontium standard solution, for ICP, in HNO₃, CRM, VHG
Standards Kationischer Standards Strontium-Standards
These solutions are intended for use as a certified reference material or calibration standard for inductively coupled plasma optical emission spectroscopy (ICP-OES), inductively coupled plasma mass spectrometry (ICP-MS), flame or furnace atomic absorption spectroscopy (AA or GFAA), x-ray fluorescence spectroscopy (XRF), and other techniques for elemental analysis.
Standards: Traceable to NIST, SRM.