Selenium standards, VHG
Lieferant: LGC Standards
VHG
PROOVHG-SPSE6-4X25EA 0 EUR
PROOVHG-SPSE6-4X25 PROOVHG-SPSE4-50 PROOVHG-SPSE6-50 PROOVHG-SPSE4-4X25 PROOVHG-SPSE4-100 PROOVHG-SPSE6-100
Selenium standards, VHG
Standards Kationischer Standards Selen-Standards
This standard solution is intended for use as a calibration and reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.
Standards: Accredited to ISO 9001 and ISO/IEC 17025. Traceable to NIST and SRM.