Standards for water pollution QC check, inorganics, VHG
Lieferant: LGC Standards
PROOVHG-QWSIN-500EA 143 EUR
PROOVHG-QWSIN-500
Standards for water pollution QC check, inorganics, VHG
Standards Umweltanalyse-Standards
This solution is intended for use as a quality control check for methods related to measurements of inorganic materials includes conductivity, alkalinity as CaCO₃, chloride, fluoride, nitrite and nitrate, sulphate, potassium, sodium, total dissolved solids and many more.
This standard has been prepared as a whole volume standard and is ready to be analysed as received. This standard is not preserved.
Standards: Traceable to NIST SRM.