Standards for water pollution QC check, total residual chlorine, VHG
Lieferant: LGC Standards
PROOVHG-QWPRCL-2EA 94 EUR
PROOVHG-QWPRCL-2
Standards for water pollution QC check, total residual chlorine, VHG
Standards Umweltanalyse-Standards
This product is intended for use as a quality control check for methods related to total residual chlorine.
This standard has been prepared as a concentrate and must be diluted prior to analysis. This concentrate is preserved with approximately 1% (v/v) hydrochloric acid (HCl).
Standards: Traceable to NIST SRM.