Standards for water pollution QC check, mercury, VHG
Lieferant: LGC Standards
PROOVHG-QWPHG-15EA 86 EUR
PROOVHG-QWPHG-15
Standards for water pollution QC check, mercury, VHG
Standards Umweltanalyse-Standards
This solution is intended for use as a quality control check for methods related to mercury.
This standard has been preserved with approximately 1% (v/v) nitric acid and 0,04% (w/v) potassium dichromate. This standard has been prepared as a concentrate and must be diluted prior to analysis.
Standards: Traceable to NIST SRM.