Standards for water pollution QC check, hexavalent chromium, VHG
Lieferant: LGC Standards
PROOVHG-QWPCR6-15EA 111 EUR
PROOVHG-QWPCR6-15
Standards for water pollution QC check, hexavalent chromium, VHG
Standards Umweltanalyse-Standards
This solution is intended for use as a quality control check for methods related to hexavalent chromium.
This standard has been prepared as a concentrate and must be diluted prior to analysis and should be analysed as soon as possible after the concentrate is diluted. This concentrate is not preserved but can be stored at room temperature.
Standards: Traceable to NIST SRM.